Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ICL
Entrez Gene ID
821726
Gene Name
isocitrate lyase
Gene Aliases
AT3G21720, isocitrate lyase
Chromosome Location
Chr.3: 7652487 - 7656053 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113067.4 NP_188809.2 4 - 5 1812 89
GenBank mRNA AK226403.1 - 4 - 5 1706 89
AK316978.1 - 4 - 5 1704 89
AY140000.1 - 4 - 5 1704 89
BT010397.1 - 4 - 5 1632 89

View Full Product Details