Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CWLP
Entrez Gene ID
821775
Gene Name
cell wall-plasma membrane linker protein
Gene Aliases
AT3G22120, cell wall-plasma membrane linker protein
Chromosome Location
Chr.3: 7794758 - 7796460 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338564.1 NP_001327425.1 1 - 1 1283 139
NM_113109.3 NP_188851.2 1 - 1 1133 139

View Full Product Details