Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIC
Entrez Gene ID
821807
Gene Name
time for coffee
Gene Aliases
AT3G22380, TIME FOR COFFEE
Chromosome Location
Chr.3: 7912905 - 7919707 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125204.1 NP_001118676.1 4 - 5 1409 113
NM_001338582.1 NP_001326717.1 4 - 5 1409 113
NM_113136.4 NP_566705.1 4 - 5 1409 113

View Full Product Details