Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOL1
Entrez Gene ID
821847
Gene Name
Tesmin/TSO1-like CXC domain-containing protein
Gene Aliases
AT3G22760
Chromosome Location
Chr.3: 8044410 - 8048263 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338596.1 NP_001325669.1 8 - 9 1547 109
NM_113175.2 NP_566717.1 8 - 9 1559 109
GenBank mRNA AY099653.1 - 8 - 9 1430 109
AY128837.1 - 8 - 9 1364 109

View Full Product Details