Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ckl10
- Entrez Gene ID
-
821915
- Gene Name
- casein kinase I-like 10
- Gene Aliases
-
AT3G23340, casein kinase I-like 10
- Chromosome Location
-
Chr.3: 8350803 - 8354021 on Build TAIR10.1
- Gene Symbol
- AT3G23350
- Entrez Gene ID
-
821916
- Gene Name
- ENTH/VHS family protein
- Gene Aliases
-
AT3G23350
- Chromosome Location
-
Chr.3: 8354077 - 8355410 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY735607.1 |
- |
5 - 6 |
414 |
75 |
View Full Product Details