Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ckl10
Entrez Gene ID
821915
Gene Name
casein kinase I-like 10
Gene Aliases
AT3G23340, casein kinase I-like 10
Chromosome Location
Chr.3: 8350803 - 8354021 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338625.1 NP_001326886.1 9 - 10 916 112
NM_113237.3 NP_188976.1 9 - 10 958 112
GenBank mRNA AY054179.1 - 9 - 10 958 112
AY074556.1 - 9 - 10 714 112
AY943849.1 - 9 - 10 714 112

View Full Product Details