Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RFR1
Entrez Gene ID
821938
Gene Name
REF4-related 1
Gene Aliases
AT3G23590, MED33A, REF4-related 1
Chromosome Location
Chr.3: 8467398 - 8473627 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113262.4 NP_189001.1 7 - 8 1904 94

View Full Product Details