Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SBP3
Entrez Gene ID
821962
Gene Name
selenium-binding protein 3
Gene Aliases
AT3G23800, selenium-binding protein 3
Chromosome Location
Chr.3: 8581446 - 8583987 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338655.1 NP_001327897.1 5 - 6 1089 111
NM_113284.3 NP_189022.1 6 - 7 1413 111
GenBank mRNA AK117618.1 - 6 - 7 1205 111
BT005102.1 - 6 - 7 1158 111

View Full Product Details