Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP60
Entrez Gene ID
821983
Gene Name
heat shock protein 60
Gene Aliases
AT3G23990, HEAT SHOCK PROTEIN 60-3B, HSP60-3B, heat shock protein 60
Chromosome Location
Chr.3: 8668772 - 8672578 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113304.5 NP_189041.1 9 - 10 1298 80
GenBank mRNA AY099594.1 - 9 - 10 1142 80
BT010398.1 - 9 - 10 1057 80

View Full Product Details