Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHR40
Entrez Gene ID
822023
Gene Name
chromatin remodeling 40
Gene Aliases
AT3G24340, chromatin remodeling 40
Chromosome Location
Chr.3: 8831935 - 8835829 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113341.2 NP_189077.1 1 - 2 1546 82

View Full Product Details