Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBC37
Entrez Gene ID
822045
Gene Name
ubiquitin-conjugating enzyme 37
Gene Aliases
AT3G24515, ubiquitin-conjugating enzyme 37
Chromosome Location
Chr.3: 8934408 - 8936298 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113362.2 NP_566751.1 2 - 3 286 102

View Full Product Details