Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLE41
Entrez Gene ID
822075
Gene Name
CLAVATA3/ESR-RELATED 41
Gene Aliases
AT3G24770, CLAVATA3/ESR-RELATED 41
Chromosome Location
Chr.3: 9046293 - 9047463 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113389.1 NP_566754.1 1 - 1 747 160

View Full Product Details