Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RALFL26
Entrez Gene ID
822109
Gene Name
ralf-like 26
Gene Aliases
AT3G25170, MJL12.13, ralf-like 26
Chromosome Location
Chr.3: 9165667 - 9166820 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338742.1 NP_001327923.1 2 - 2 282 88
NM_001338743.1 NP_001327922.1 2 - 2 301 88
NM_113422.4 NP_566761.1 1 - 1 514 88

View Full Product Details