Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP82G1
Entrez Gene ID
822110
Gene Name
cytochrome P450, family 82, subfamily G, polypeptide 1
Gene Aliases
AT3G25180, cytochrome P450, family 82, polypeptide 1, subfamily G
Chromosome Location
Chr.3: 9167138 - 9169301 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113423.4 NP_189154.1 1 - 2 960 73
NM_202630.2 NP_974359.1 1 - 2 960 73

View Full Product Details