Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ROF1
Entrez Gene ID
822117
Gene Name
rotamase FKBP 1
Gene Aliases
AT3G25230, ATFKBP62, FK506 BINDING PROTEIN 62, FKBP62, rotamase FKBP 1
Chromosome Location
Chr.3: 9188096 - 9191630 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125223.1 NP_001118695.1 5 - 6 768 97
NM_113429.5 NP_189160.3 5 - 6 853 97

View Full Product Details