Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AFH1
Entrez Gene ID
822136
Gene Name
formin homology 1
Gene Aliases
AHF1, ARABIDOPSIS THALIANA FORMIN HOMOLOGY 1, AT3G25500, ATFH1, FH1, FORMIN HOMOLOGY 1, FORMIN-LIKE PROTEIN AHF1, formin homology 1
Chromosome Location
Chr.3: 9251093 - 9255083 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113446.5 NP_189177.1 1 - 2 2129 82
GenBank mRNA AF174427.1 - 1 - 2 1904 82
AY080793.1 - 1 - 2 1974 82
AY142579.1 - 1 - 2 1872 82

View Full Product Details