Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP1B
Entrez Gene ID
822165
Gene Name
methionine aminopeptidase 1C
Gene Aliases
AT3G25740, MAP1C, METHIONINE AMINOPEPTIDASE 1C, methionine aminopeptidase 1C
Chromosome Location
Chr.3: 9397784 - 9399805 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113473.3 NP_189202.1 5 - 6 603 108
GenBank mRNA BT024875.1 - 5 - 6 581 108

View Full Product Details