Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGS1
Entrez Gene ID
822207
Gene Name
REGULATOR OF G-PROTEIN SIGNALING 1
Gene Aliases
AT3G26090, ATRGS1, REGULATOR OF G-PROTEIN SIGNALING 1
Chromosome Location
Chr.3: 9532613 - 9535629 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338780.1 NP_001327634.1 7 - 8 1221 130
NM_113514.4 NP_189238.2 7 - 8 1221 130
GenBank mRNA AY150429.1 - 7 - 8 951 130

View Full Product Details