Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP71B26
Entrez Gene ID
822232
Gene Name
cytochrome P450, family 71, subfamily B, polypeptide 26
Gene Aliases
''cytochrome P450, AT3G26290, cytochrome P450, family 71, polypeptide 26, polypeptide 26'', subfamily B
Chromosome Location
Chr.3: 9632608 - 9634822 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338796.1 NP_001327178.1 1 - 2 692 113
NM_113536.3 NP_189260.1 1 - 2 340 113
GenBank mRNA AY128393.1 - 1 - 2 339 113
BT008871.1 - 1 - 2 309 113

View Full Product Details