Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAPA
Entrez Gene ID
822277
Gene Name
glyceraldehyde 3-phosphate dehydrogenase A subunit
Gene Aliases
AT3G26650, GAPA-1, GLYCERALDEHYDE 3-PHOSPHATE DEHYDROGENASE A SUBUNIT, GLYCERALDEHYDE 3-PHOSPHATE DEHYDROGENASE A SUBUNIT 1, GLYCERALDEHYDE-3-PHOSPHATE DEHYDROGENASE A SUBUNIT, glyceraldehyde 3-phosphate dehydrogenase A subunit
Chromosome Location
Chr.3: 9794862 - 9797045 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113576.4 NP_566796.2 3 - 4 811 67
GenBank mRNA AF428431.1 - 3 - 4 494 67
AY058107.1 - 3 - 4 488 67
AY058140.1 - 3 - 4 494 67
AY075637.1 - 3 - 4 496 67
AY142053.1 - 3 - 4 447 67

View Full Product Details