Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MUB4
Entrez Gene ID
822315
Gene Name
membrane-anchored ubiquitin-fold protein 4 precursor
Gene Aliases
AT3G26980, membrane-anchored ubiquitin-fold protein 4 precursor
Chromosome Location
Chr.3: 9945756 - 9948084 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113612.4 NP_189334.2 2 - 3 489 131
GenBank mRNA AY045950.1 - 2 - 3 444 131
AY079339.1 - 1 - 2 124 131

View Full Product Details