Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TCP20
- Entrez Gene ID
-
822318
- Gene Name
- TEOSINTE BRANCHED 1, cycloidea, PCF (TCP)-domain family protein 20
- Gene Aliases
-
ARABIDOPSIS THALIANA TEOSINTE BRANCHED 1, AT-TCP20, AT3G27010, ATTCP20, CYCLOIDEA, PCF (TCP)-DOMAIN FAMILY PROTEIN 20, PCF (TCP)-domain family protein 20, PCF1, TEOSINTE BRANCHED 1, cycloidea
- Chromosome Location
-
Chr.3: 9957376 - 9958945 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_113615.4 |
NP_189337.1 |
1 - 2 |
1300 |
113 |
View Full Product Details