Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YSL6
Entrez Gene ID
822319
Gene Name
YELLOW STRIPE like 6
Gene Aliases
AT3G27020, AtYSL6, YELLOW STRIPE like 6
Chromosome Location
Chr.3: 9961277 - 9964737 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113616.3 NP_566806.1 6 - 7 1363 76

View Full Product Details