Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIPL2
Entrez Gene ID
836384
Gene Name
hipl2 protein precursor
Gene Aliases
AT5G62630, MRG21.5, MRG21_5, hipl2 protein precursor
Chromosome Location
Chr.5: 25143518 - 25146587 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125658.3 NP_201069.1 4 - 5 1446 124

View Full Product Details