Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDPK2
Entrez Gene ID
836451
Gene Name
nucleoside diphosphate kinase 2
Gene Aliases
ARABIDOPSIS NUCLEOSIDE DIPHOSPHATE KINASE 2, AT5G63310, ATNDPK2, MDC12.28, MDC12_28, NDP KINASE 1A, NDPK IA, NDPK IA IA, NDPK1A, NUCLEOSIDE DIPHOSPHATE KINASE IA, nucleoside diphosphate kinase 2
Chromosome Location
Chr.5: 25371904 - 25373904 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125726.4 NP_568970.2 5 - 6 648 93
GenBank mRNA AJ012758.1 - 5 - 6 582 93
AK227190.1 - 5 - 6 603 93
AY057612.1 - 5 - 6 601 93
AY065291.1 - 5 - 6 599 93
AY117366.1 - 5 - 6 582 93

View Full Product Details