Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RMI1
Entrez Gene ID
836473
Gene Name
recQ-mediated instability protein (DUF1767)
Gene Aliases
AT5G63540, ATRMI1, BLAP75, MLE2.17, MLE2_17, RECQ MEDIATED INSTABILITY 1
Chromosome Location
Chr.5: 25440783 - 25444471 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126014.1 NP_001119486.1 5 - 6 1171 75
NM_125749.5 NP_201159.2 4 - 5 1105 75
GenBank mRNA AY735746.1 - 4 - 5 1070 75
AY954880.1 - 4 - 5 1007 75

View Full Product Details