Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHA1
Entrez Gene ID
836498
Gene Name
RING/FYVE/PHD zinc finger superfamily protein
Gene Aliases
AT5G63780, MBK5.26, MBK5_26, shoot apical meristem arrest 1
Chromosome Location
Chr.5: 25523827 - 25526593 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345611.1 NP_001331890.1 3 - 4 1992 79
NM_125773.4 NP_201183.2 3 - 4 1445 79
GenBank mRNA BT002521.1 - 3 - 4 1428 79
BT008456.1 - 2 - 3 987 79

View Full Product Details