Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MUM2
- Entrez Gene ID
-
836500
- Gene Name
- Glycosyl hydrolase family 35 protein
- Gene Aliases
-
AT5G63800, BGAL6, MBK5.28, MBK5_28, MUCILAGE-MODIFIED 2, beta-galactosidase 6
- Chromosome Location
-
Chr.5: 25530055 - 25535889 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125775.4 |
NP_568978.2 |
1 - 2 |
453 |
79 |
View Full Product Details