Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAP4
Entrez Gene ID
836505
Gene Name
amino acid permease 4
Gene Aliases
AT5G63850, MGI19.6, MGI19_6, amino acid permease 4
Chromosome Location
Chr.5: 25550937 - 25553656 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125780.3 NP_201190.1 5 - 6 1018 64

View Full Product Details