Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAL2
Entrez Gene ID
836521
Gene Name
Inositol monophosphatase family protein
Gene Aliases
AT5G64000, ATSAL2, MBM17.10, MBM17_10
Chromosome Location
Chr.5: 25616625 - 25618631 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125796.5 NP_201205.1 2 - 3 186 100
GenBank mRNA AY070383.1 - 2 - 3 173 100
AY096575.1 - 2 - 3 147 100
Z83312.1 - - 183 100

View Full Product Details