Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERS
Entrez Gene ID
836526
Gene Name
glutamate tRNA synthetase
Gene Aliases
AT5G64050, ATERS, ETHYLENE RESPONSE SENSOR, OVA3, OVULE ABORTION 3, glutamate tRNA synthetase
Chromosome Location
Chr.5: 25629832 - 25633209 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125801.4 NP_201210.1 7 - 8 1050 147
GenBank mRNA AF241841.1 - 7 - 8 1005 147

View Full Product Details