Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXL2
Entrez Gene ID
836547
Gene Name
EXORDIUM like 2
Gene Aliases
AT5G64260, EXORDIUM like 2, MSJ1_10
Chromosome Location
Chr.5: 25703649 - 25705339 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125822.3 NP_201231.1 1 - 1 1423 75
GenBank mRNA AF326891.1 - 1 - 1 1143 75
AF380638.1 - 1 - 1 1143 75

View Full Product Details