Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GCH
- Entrez Gene ID
-
836551
- Gene Name
- GTP cyclohydrolase II
- Gene Aliases
-
ARABIDOPSIS THALIANA GTP CYCLOHYDROLASE II, AT5G64300, ATGCH, ATRIBA1, Arabidopsis thaliana riboflavin A1, GTP CYCLOHYDROLASE II, GTP cyclohydrolase II, RED FLUORESCENT IN DARKNESS 1, RFD1, RIBA1, riboflavin A1
- Chromosome Location
-
Chr.5: 25718008 - 25721166 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125826.5 |
NP_201235.4 |
6 - 7 |
1848 |
122 |
View Full Product Details