Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPN5B
Entrez Gene ID
836597
Gene Name
regulatory particle non-ATPase subunit 5B
Gene Aliases
AT5G64760, MVP7.9, MVP7_9, regulatory particle non-ATPase subunit 5B
Chromosome Location
Chr.5: 25893780 - 25896586 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037065.2 NP_001032142.1 6 - 7 774 101
NM_001345648.1 NP_001332553.1 5 - 6 877 101
NM_125872.4 NP_568994.2 6 - 7 805 101
GenBank mRNA AY062093.2 - 6 - 7 780 101
AY062113.1 - 6 - 7 780 101
AY124872.1 - 5 - 6 664 101
AY230833.1 - 5 - 6 667 101

View Full Product Details