Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACX2
Entrez Gene ID
836635
Gene Name
acyl-CoA oxidase 2
Gene Aliases
ACYL-COA OXIDASE 2, AT5G65110, ATACX2, MQN23.4, MQN23_4, acyl-CoA oxidase 2
Chromosome Location
Chr.5: 26009521 - 26012684 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037068.1 NP_001032145.1 5 - 6 1834 64
NM_125910.6 NP_201316.1 5 - 6 1999 64

View Full Product Details