Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSL2
Entrez Gene ID
836700
Gene Name
HAESA-like 2
Gene Aliases
AT5G65710, HAESA-like 2, MPA24.6, MPA24_6
Chromosome Location
Chr.5: 26292372 - 26295687 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125968.3 NP_201372.2 1 - 2 2603 94
GenBank mRNA FJ708817.1 - 1 - 2 2603 94

View Full Product Details