Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACR1
Entrez Gene ID
836718
Gene Name
ACT domain repeat 1
Gene Aliases
ACT domain repeat 1, AT5G65890, K14B20.6, K14B20_6
Chromosome Location
Chr.5: 26354853 - 26357905 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085334.1 NP_001078803.1 4 - 5 512 72
NM_001345724.1 NP_001330475.1 5 - 6 676 72
NM_001345725.1 NP_001330476.1 2 - 3 316 72
NM_001345726.1 NP_001330474.1 1 - 2 145 72
NM_125986.3 NP_201390.1 5 - 6 537 72
GenBank mRNA AF360256.1 - 5 - 6 502 72
AF528058.1 - 4 - 5 349 72
AY040070.1 - 4 - 5 298 72

View Full Product Details