Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATBETAFRUCT4
Entrez Gene ID
837777
Gene Name
Glycosyl hydrolases family 32 protein
Gene Aliases
AT1G12240, AtFRUCT4, AtVI2, FRUCT4, T28K15.3, T28K15_3, VAC-INV, VACUOLAR INVERTASE, VI2, fructosidase 4, vacuolar invertase 2
Chromosome Location
Chr.1: 4153641 - 4157653 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101096.3 NP_563901.1 5 - 6 1745 132
GenBank mRNA AY046009.1 - 5 - 6 1726 132
AY142666.1 - 5 - 6 1687 132

View Full Product Details