Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARK3
Entrez Gene ID
837799
Gene Name
armadillo repeat kinesin 3
Gene Aliases
AT1G12430, Arabidopsis thaliana KINESIN Ungrouped clade, AtKINUa, PAK, armadillo repeat kinesin 3, gene A, phosphatidic acid kinase
Chromosome Location
Chr.1: 4233717 - 4238795 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198043.1 NP_001184972.1 4 - 5 1039 122
NM_101115.3 NP_563908.1 4 - 5 1070 122
GenBank mRNA AB290930.1 - 4 - 5 844 122
AF159052.1 - 4 - 5 1034 122

View Full Product Details