Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCS
Entrez Gene ID
837808
Gene Name
copper chaperone for SOD1
Gene Aliases
AT1G12520, ATCCS, COPPER/ZINC SUPEROXIDE DISMUTASE COPPER CHAPERONE, F5O11.26, F5O11_26, copper chaperone for SOD1
Chromosome Location
Chr.1: 4267074 - 4268982 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035953.2 NP_001031030.1 2 - 3 280 98
NM_101123.4 NP_563910.2 2 - 3 348 98
GenBank mRNA AK175663.1 - 2 - 3 267 98
AK221656.1 - 2 - 3 268 98

View Full Product Details