Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAPA-2
Entrez Gene ID
837848
Gene Name
glyceraldehyde 3-phosphate dehydrogenase A subunit 2
Gene Aliases
AT1G12900, F13K23.15, F13K23_15, glyceraldehyde 3-phosphate dehydrogenase A subunit 2
Chromosome Location
Chr.1: 4392438 - 4394454 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084060.1 NP_001077529.1 4 - 5 820 88
NM_001123804.1 NP_001117276.1 4 - 5 740 88
NM_001198048.1 NP_001184977.1 4 - 5 856 88
NM_001332055.1 NP_001321059.1 4 - 5 870 88
NM_101161.3 NP_172750.1 4 - 5 816 88
GenBank mRNA BT008637.1 - 4 - 5 727 88
BT015867.1 - 4 - 5 699 88

View Full Product Details