Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ORP1D
Entrez Gene ID
837875
Gene Name
OSBP(oxysterol binding protein)-related protein 1D
Gene Aliases
AT1G13170, F3F19.19, F3F19_19, OSBP(oxysterol binding protein)-related protein 1D
Chromosome Location
Chr.1: 4488533 - 4492667 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198051.1 NP_001184980.1 2 - 3 1213 98
NM_101187.3 NP_172776.1 2 - 3 1293 98
GenBank mRNA AF462814.1 - 2 - 3 1209 98

View Full Product Details