Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIS1
Entrez Gene ID
837876
Gene Name
Actin-like ATPase superfamily protein
Gene Aliases
ACTIN-RELATED PROTEIN 3, ARABIDOPSIS THALIANA ACTIN-RELATED PROTEIN 3, ARP3, AT1G13180, ATARP3, DISTORTED TRICHOMES 1, F3F19.20, F3F19_20
Chromosome Location
Chr.1: 4494924 - 4498469 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101188.3 NP_172777.1 4 - 5 1215 117
GenBank mRNA AF507911.1 - 4 - 5 745 117
AY093149.1 - 4 - 5 1111 117
BT006584.1 - 4 - 5 745 117

View Full Product Details