Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAG13
Entrez Gene ID
837919
Gene Name
LAG1 longevity assurance-like protein
Gene Aliases
AT1G13580, F13B4.7, F13B4_7, LAG One Homologue 3, LAG1 longevity assurance homolog 3, LOH3
Chromosome Location
Chr.1: 4644521 - 4647142 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035960.2 NP_001031037.1 2 - 3 737 74
NM_001198056.2 NP_001184985.1 3 - 4 650 74
NM_101228.2 NP_172815.2 2 - 3 448 74
GenBank mRNA BT010465.1 - 2 - 3 360 74

View Full Product Details