Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKS2
Entrez Gene ID
837989
Gene Name
phytochrome kinase substrate 2
Gene Aliases
AT1G14280, F14L17.4, F14L17_4, phytochrome kinase substrate 2
Chromosome Location
Chr.1: 4877506 - 4879307 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101294.3 NP_172880.1 1 - 1 1463 90
GenBank mRNA AY054676.1 - 1 - 1 1332 90

View Full Product Details