Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AL7
Entrez Gene ID
838013
Gene Name
alfin-like 7
Gene Aliases
AT1G14510, AtAL7, F14L17.29, F14L17_29, alfin-like 7
Chromosome Location
Chr.1: 4961594 - 4964515 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101318.4 NP_172903.1 4 - 5 846 79
GenBank mRNA AK228067.1 - 4 - 5 687 79
AY088087.1 - 4 - 5 756 79
BT006199.1 - 4 - 5 485 79

View Full Product Details