Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DECOY
Entrez Gene ID
838024
Gene Name
decoy
Gene Aliases
AT1G14620, T5E21.12, T5E21_12
Chromosome Location
Chr.1: 5014675 - 5016708 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101329.4 NP_172914.1 5 - 6 757 78
GenBank mRNA AY127001.1 - 5 - 6 653 78
BT001031.1 - 5 - 6 550 78

View Full Product Details