Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTAC6
Entrez Gene ID
838761
Gene Name
plastid transcriptionally active 6
Gene Aliases
AT1G21600, F24J8.20, plastid transcriptionally active 6
Chromosome Location
Chr.1: 7570845 - 7573847 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035999.3 NP_001031076.1 5 - 6 928 86
NM_102009.3 NP_564144.1 5 - 6 929 86
GenBank mRNA AF324715.2 - 5 - 6 884 86
AF326894.1 - 5 - 6 884 86
AF446886.1 - 5 - 6 864 86
AK317001.1 - 5 - 6 864 86
AY052666.1 - 5 - 6 928 86
AY113873.1 - 5 - 6 864 86

View Full Product Details