Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NdhO
Entrez Gene ID
843827
Gene Name
NAD(P)H:plastoquinone dehydrogenase complex subunit O
Gene Aliases
AT1G74880, F25A4.15, F25A4_15, NAD(P)H:plastoquinone dehydrogenase complex subunit O, NADH dehydrogenase-like complex ), NDH-O
Chromosome Location
Chr.1: 28129828 - 28131077 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106146.5 NP_565093.1 4 - 5 447 124
GenBank mRNA AF344320.1 - 4 - 5 390 124
AF410289.1 - 4 - 5 447 124
AY143808.1 - 4 - 5 390 124

View Full Product Details