The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198470.1 | NP_001185399.1 | 6 - 7 | 847 | 64 |
NM_106150.4 | NP_565094.1 | 6 - 7 | 955 | 64 | |
GenBank mRNA | AY087395.1 | - | 6 - 7 | 862 | 64 |
AY093071.1 | - | 6 - 7 | 820 | 64 | |
BT008872.1 | - | 6 - 7 | 691 | 64 |